3-Carene, 90%, stabilized
3-Carene, 90%, stabilized
3-Carene, 90%, stabilized
3-Carene, 90%, stabilized
Thermo Scientific Chemicals
3-Carene, 90%, stabilized
CAS: 13466-78-9 | C10H16 | 136.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
Packaging:
Glass bottle
Catalog number FSA367960250
Price (MYR)
278.00
ML
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
278.00
ML
Add to cart
Specifications
Chemical Name or Material3-Carene
CAS13466-78-9
CAS Min %88.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0