2-Bromoethyl ethyl ether, 90%
2-Bromoethyl ethyl ether, 90%
2-Bromoethyl ethyl ether, 90%
2-Bromoethyl ethyl ether, 90%
Thermo Scientific Chemicals
2-Bromoethyl ethyl ether, 90%
CAS: 592-55-2 | C4H9BrO | 153.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA367401000
Price (MYR)
1,624.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,624.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Bromoethyl ethyl ether
CAS592-55-2
CAS Min %88.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0