3-Hexylthiophene, 98%
3-Hexylthiophene, 98%
3-Hexylthiophene, 98%
3-Hexylthiophene, 98%
Thermo Scientific Chemicals
3-Hexylthiophene, 98%
CAS: 1693-86-3 | C10H16S | 168.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA367380250
Price (MYR)
2,222.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,222.00
EA
Add to cart
Specifications
Chemical Name or Material3-Hexylthiophene
CAS1693-86-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0