4-Hydroxypiperidine, 99+%
4-Hydroxypiperidine, 99+%
4-Hydroxypiperidine, 99+%
4-Hydroxypiperidine, 99+%
Thermo Scientific Chemicals
4-Hydroxypiperidine, 99+%
CAS: 5382-16-1 | C5H11NO | 101.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA366510250
Price (MYR)
485.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
485.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Hydroxypiperidine
Name Note99+%
CAS5382-16-1
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger