Tetrabutylammonium hydroxide, 40 wt.% solution in methanol
Tetrabutylammonium hydroxide, 40 wt.% solution in methanol
Tetrabutylammonium hydroxide, 40 wt.% solution in methanol
Tetrabutylammonium hydroxide, 40 wt.% solution in methanol
Thermo Scientific Chemicals
Tetrabutylammonium hydroxide, 40 wt.% solution in methanol
CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA366325000
Price (MYR)
1,978.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,978.00
BOT
Add to cart
Chemical Identifiers
CAS2052-49-5, 67-56-1
Specifications
Chemical Name or MaterialTetrabutylammonium hydroxide
Name Note40 wt.% solution in methanol
CAS2052-49-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
View more
Phase transfer catalyst
RUO – Research Use Only