Ethanesulfonic acid, 95%
Ethanesulfonic acid, 95%
Ethanesulfonic acid, 95%
Ethanesulfonic acid, 95%
Thermo Scientific Chemicals
Ethanesulfonic acid, 95%
CAS: 594-45-6 | C2H5O3S | 109.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA366041000
Price (MYR)
1,626.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,626.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthanesulfonic acid
Name Note95%
CAS594-45-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement