Dodecane, mixture of isomers
Dodecane, mixture of isomers
Dodecane, mixture of isomers
Dodecane, mixture of isomers
Thermo Scientific Chemicals
Dodecane, mixture of isomers
Dodecane, C12H26, CAS Number-13475-82-6, heptane,2,2,4,6,6-pentamethyl, 2,2,4,6,6-pentamethyl-heptane, heptane, 2,2,4,6,6-pentamethyl, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, permethyl 99a, 1L, 170.0 deg.C to 195.0 deg.C, CHEBI:77509, Undesignated, 0.7510g/mL, 236-757-, 44 deg.C, 170.34 | CAS: 13475-82-6 | C12H26 | 170.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
Packaging:
View all
Metal drum
Catalog number FSA365770250
Price (MYR)
3,512.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Price (MYR)
3,512.00
EA
Add to cart
Specifications
Chemical Name or MaterialDodecane
CAS13475-82-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o