Coconut oil, pure, refined
Coconut oil, pure, refined
Thermo Scientific Chemicals
Coconut oil, pure, refined
CAS: 8001-31-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA365475000
Price (MYR)
353.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
353.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCoconut oil
Name NoteRefined
Acidity0.5mg KOH/g max.
CAS8001-31-8
Melting Point23.0°C to 26.0°C