L(+)-Arabinose, 99%, for biochemistry
L(+)-Arabinose, 99%, for biochemistry
L(+)-Arabinose, 99%, for biochemistry
L(+)-Arabinose, 99%, for biochemistry
Thermo Scientific Chemicals
L(+)-Arabinose, 99%, for biochemistry
CAS: 87-72-9 | C5H10O5 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number FSA365185000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL(+)-Arabinose
CAS87-72-9
Melting Point155.0°C to 160.0°C
EINECS Number201-767-6
Ignition Residue0.3% max