n-Hexane, 97%, Extra Dry over Molecular Sieve, AcroSeal™
n-Hexane, 97%, Extra Dry over Molecular Sieve, AcroSeal™
n-Hexane, 97%, Extra Dry over Molecular Sieve, AcroSeal™
n-Hexane, 97%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals
n-Hexane, 97%, Extra Dry over Molecular Sieve, AcroSeal™
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA364370010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Hexane
Melting Point-95.0°C
Name NoteOver Molecular Sieves
CAS110-54-3
CAS Min %96.0