Di-μ-chlorobis(p-cymene)chlororuthenium(II), 98%
Di-μ-chlorobis(p-cymene)chlororuthenium(II), 98%
Di-μ-chlorobis(p-cymene)chlororuthenium(II), 98%
Di-μ-chlorobis(p-cymene)chlororuthenium(II), 98%
Thermo Scientific Chemicals
Di-μ-chlorobis(p-cymene)chlororuthenium(II), 98%
CAS: 52462-29-0 | C20H28Cl4Ru2 | 612.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA363290010
Price (MYR)
540.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
540.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDi-μ-chlorobis(p-cymene)chlororuthenium(II)
CAS52462-29-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc