4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA359810050
Price (MYR)
1,312.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,312.00
EA
Add to cart
Specifications
Chemical Name or Material4-Vinylbenzeneboronic acid
CAS2156-04-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0