4-Chloro-1-butanol, 85%, balance THF and HCl
4-Chloro-1-butanol, 85%, balance THF and HCl
4-Chloro-1-butanol, 85%, balance THF and HCl
4-Chloro-1-butanol, 85%, balance THF and HCl
Thermo Scientific Chemicals
4-Chloro-1-butanol, 85%, balance THF and HCl
CAS: 928-51-8 | C4H9ClO | 108.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA356310100
Price (MYR)
336.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
336.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Chloro-1-butanol
Name NoteBalance THF and HCl
CAS928-51-8
CAS Min %80.0
Health Hazard 1GHS Signal Word: Warning