Cyclopentanecarbonyl chloride, 98%
Cyclopentanecarbonyl chloride, 98%
Cyclopentanecarbonyl chloride, 98%
Cyclopentanecarbonyl chloride, 98%
Thermo Scientific Chemicals
Cyclopentanecarbonyl chloride, 98%
CAS: 4524-93-0 | C6H9ClO | 132.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA354370250
Price (MYR)
1,323.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,323.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCyclopentanecarbonyl chloride
CAS4524-93-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi