1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
Thermo Scientific Chemicals
1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
CAS: 304680-35-1 | C10H19F6N2P | 312.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Plastic bottle
Catalog number FSA354181000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Hexyl-3-methylimidazolium hexafluorophosphate
CAS304680-35-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Ionic liquid
RUO – Research Use Only