Dibutyl maleate, 97%
Dibutyl maleate, 97%
Dibutyl maleate, 97%
Dibutyl maleate, 97%
Thermo Scientific Chemicals
Dibutyl maleate, 97%
CAS: 105-76-0 | C12H20O4 | 228.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA353102500
Price (MYR)
268.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
268.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDibutyl maleate
Name Note97%
CAS105-76-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.