Hydrocortisone, 98%
Hydrocortisone, 98%
Hydrocortisone, 98%
Thermo Scientific Chemicals
Hydrocortisone, 98%
Hydrocortisone, CAS # 50-23-7, is the pharmaceutical formulation of the hormone, cortisol, and is commonly administered as an immunosuppressive drug.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA352451000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrocortisone
CAS50-23-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
View more
General description
• Hydrocortisone is the synthetic formulation of the naturally occurring hormone, cortisol
• It is available as a white crystalline powder soluble in water, DMSO, and ethanol
• It is stable at room temperature but sensitive to light, and reactive in the presence of strong oxidizing agents
Applications
• Hydrocortisone works as an agonist to glucocorticoid receptors, thereby suppressing the inflammatory response of the immune system
• It’s administration induces gluconeogenesis, thus increasing endogenous blood sugar levels
• Research indicates a role for hydrocortisone in the metabolism of fat, protein, and carbohydrates
RUO – Research Use Only
General References:
  1. Ramamoorthy, S. and Cidlowski, J. A. Corticosteroids-Mechanisms of Action in Health and Disease. Rheum Dis Clin North Am. 2016, 42(1): 15–31.
  2. Spireas, S.; Sadu, S.; and Grover, R. In vitro release evaluation of hydrocortisone liquisolid tablets. Journal of Pharmaceutical Sciences. 1998, 87(7), 867-872.
  3. Oji, N. and Shreeve, W. W. Acute effects of hydrocortisone on gluconeogenesis from 14C- and 3H-labeled substrates in intact rats Endocrinology. 1967, 80(6), 1062-8.
  4. Dandona. P; Thusu, K.; Hafeez, R.; Abdel-Rahman, E. Effect of hydrocortisone on oxygen free radical generation by mononuclear cells. Metabolism. 1998, 47(7), 788-791.