Pentaethylene glycol, 98%
Pentaethylene glycol, 98%
Pentaethylene glycol, 98%
Pentaethylene glycol, 98%
Thermo Scientific Chemicals
Pentaethylene glycol, 98%
CAS: 4792-15-8 | C10H22O6 | 238.28 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3507300505 gGlass bottle
Catalog number FSA350730050
Price (MYR)
385.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
385.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPentaethylene glycol
Name Note98%
CAS4792-15-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: