2,3-Diamino-5-bromopyridine, 97%
2,3-Diamino-5-bromopyridine, 97%
2,3-Diamino-5-bromopyridine, 97%
2,3-Diamino-5-bromopyridine, 97%
Thermo Scientific Chemicals
2,3-Diamino-5-bromopyridine, 97%
CAS: 38875-53-5 | C5H7BrN3 | 189.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA350330050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 3-Diamino-5-bromopyridine
CAS38875-53-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0