Di-tert-butyl peroxide, 99%
Di-tert-butyl peroxide, 99%
Di-tert-butyl peroxide, 99%
Di-tert-butyl peroxide, 99%
Thermo Scientific Chemicals
Di-tert-butyl peroxide, 99%
CAS: 110-05-4 | C8H18O2 | 146.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA349931000
Price (MYR)
343.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
343.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDi-tert-butyl peroxide
CAS110-05-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Heating may cause a fire.
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get me
View more
Radical initiators
RUO – Research Use Only