Dilauroyl peroxide, 99%
Dilauroyl peroxide, 99%
Dilauroyl peroxide, 99%
Dilauroyl peroxide, 99%
Thermo Scientific Chemicals
Dilauroyl peroxide, 99%
CAS: 105-74-8 | C24H46O4 | 398.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA349741000
Price (MYR)
446.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
446.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDilauroyl peroxide
Name Note99%
CAS105-74-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Heating may cause a fire.
View more
Radical initiators
RUO – Research Use Only