2',7'-Dichlorofluorescein 3',6'-diacetate, 97%
2',7'-Dichlorofluorescein 3',6'-diacetate, 97%
2',7'-Dichlorofluorescein 3',6'-diacetate, 97%
2',7'-Dichlorofluorescein 3',6'-diacetate, 97%
Thermo Scientific Chemicals
2',7'-Dichlorofluorescein 3',6'-diacetate, 97%
CAS: 2044-85-1 | C24H14Cl2O7 | 485.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA347250050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2', 7'-Dichlorofluorescein 3', 6'-diacetate
Melting Point239°C to 246°C
CAS2044-85-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger