Coumarin-3-carboxylic acid, 98%
Coumarin-3-carboxylic acid, 98%
Coumarin-3-carboxylic acid, 98%
Coumarin-3-carboxylic acid, 98%
Thermo Scientific Chemicals
Coumarin-3-carboxylic acid, 98%
CAS: 531-81-7 | C10H6O4 | 190.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA346760250
Price (MYR)
427.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
427.00
EA
Add to cart
Specifications
Chemical Name or MaterialCoumarin-3-carboxylic acid
CAS531-81-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic if swallowed.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.