3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech.
3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech.
3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech.
3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech.
3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech.
Thermo Scientific Chemicals
3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech.
CAS: 35141-30-1 | C10H27N3O3Si | 265.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA346171000
Price (MYR)
545.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
545.00
BOT
Add to cart
Specifications
Chemical Name or Material3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane
CAS35141-30-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if inhaled.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lens