Carbamazepine, 98%
Carbamazepine, 98%
Carbamazepine, 98%
Thermo Scientific Chemicals
Carbamazepine, 98%
Carbamazepine (Carbamazepen), CAS # 298-46-4, is an aromatic ring of the dibenzoazepine class that acts as a sodium channel blocker. | CAS: 298-46-4 | C15H12N2O | 236.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA344360250
Price (MYR)
1,625.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,625.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCarbamazepine
Name Note98%
CAS298-46-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
View more
General Description
  • Carbamazepine is an aromatic ring of the dibenzoazepine class that acts as a sodium channel blocker
  • It is a tricyclic antidepressant-like drug that reduces post-tetanic potentiation
Applications
  • Carbamazepine can be used as a sodium channel blocker and as a glutamate-transporter agonist to reduce neural activity and produce inhibitory effects in the brain
  • Carbamazepine is useful in the research of pain, epilepsy, and manic depression
RUO – Research Use Only
General References:
  1. Ben-Menachem, E.; Grebe, H.P.; Terada, K.; Jensen, L.; Li, T.; De Backer, M.; Steiniger-Brach, B.; Gasalla, T.; Brock, M.; Biton, V. Long-term safety and efficacy of lacosamide and controlled-release carbamazepine monotherapy in patients with newly diagnosed epilepsy. Epilepsia. 2019, 60(12), 2437-2447. Erratum in: Epilepsia. 2021, 62(4), 1039.
  2. Schmitz, B.; Dimova, S.; Zhang, Y.; Chellun, D.; De Backer, M.; Gasalla, T. Tolerability and efficacy of lacosamide and controlled-release carbamazepine monotherapy in patients with newly diagnosed epilepsy and concomitant psychiatric conditions: Post hoc analysis of a prospective, randomized, double-blind trial. Epilepsy Res. 2020, 159, 106220.