Formononetin, 98%
Formononetin, 98%
Formononetin, 98%
Thermo Scientific Chemicals
Formononetin, 98%
CAS: 485-72-3 | C16H12O4 | 268.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3424800101 gGlass bottle
Catalog number FSA342480010
Price (MYR)
751.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
751.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFormononetin
Name Note99%
CAS485-72-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: