Nicotinoyl chloride hydrochloride, 97%
Nicotinoyl chloride hydrochloride, 97%
Nicotinoyl chloride hydrochloride, 97%
Nicotinoyl chloride hydrochloride, 97%
Thermo Scientific Chemicals
Nicotinoyl chloride hydrochloride, 97%
CAS: 20260-53-1 | C6H4ClNO·HCl | 178.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA338410250
Price (MYR)
633.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
633.00
BOT
Add to cart
Specifications
Chemical Name or MaterialNicotinoyl chloride hydrochloride
Name Note97%
CAS20260-53-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger