4-Nitrophenyl-beta-D-glucuronide, 99+%
4-Nitrophenyl-beta-D-glucuronide, 99+%
4-Nitrophenyl-beta-D-glucuronide, 99+%
4-Nitrophenyl-beta-D-glucuronide, 99+%
Thermo Scientific Chemicals
4-Nitrophenyl-beta-D-glucuronide, 99+%
CAS: 10344-94-2 | C12H12NO9 | 314.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
50 mg
1 g
Packaging:
Glass bottle
Catalog number FSA336792500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitrophenyl-β-D-glucuronide
CAS10344-94-2
CAS Min %99.0
Health Hazard 1MOISTURE SENSITIVE
CAS Max %100.0