Check Order Status
3-Methylpicolinic acid, 98%, Thermo Scientific™
Element Search
Structure Search
3-Methylpicolinic acid, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
Packaging:
Glass bottle
Catalog number
FSA335960050
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
3-Methylpicolinic acid
CAS
4021-07-2
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0