1-Allylimidazole, 97%
1-Allylimidazole, 97%
1-Allylimidazole, 97%
1-Allylimidazole, 97%
Thermo Scientific Chemicals
1-Allylimidazole, 97%
CAS: 31410-01-2 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA335171000
Price (MYR)
632.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
632.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Allylimidazole
Name Note97%
CAS31410-01-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger