4-Isopropylbenzaldehyde, 98%
4-Isopropylbenzaldehyde, 98%
4-Isopropylbenzaldehyde, 98%
4-Isopropylbenzaldehyde, 98%
Thermo Scientific Chemicals
4-Isopropylbenzaldehyde, 98%
CAS: 122-03-2 | C10H12O | 148.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA334881000
Price (MYR)
321.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
321.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Isopropylbenzaldehyde
Name Note98%
CAS122-03-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning