(1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%
(1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%
(1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%
(1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%
(1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%
CAS: 716-61-0 | C9H13N2O4 | 213.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA334790250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(1R, 2R)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol
Name Note96%
CAS716-61-0
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning