Polysorbate 85
Polysorbate 85
Polysorbate 85
Thermo Scientific Chemicals
Polysorbate 85
CAS: 9005-70-3 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7 | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA334152500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPolysorbate 85
ColorColorless
Acid Value2mg KOH/g max.
CAS9005-70-3
Physical FormLiquid