2-Mercaptobenzothiazole, 97%
2-Mercaptobenzothiazole, 97%
2-Mercaptobenzothiazole, 97%
2-Mercaptobenzothiazole, 97%
Thermo Scientific Chemicals
2-Mercaptobenzothiazole, 97%
CAS: 149-30-4 | C7H5NS2 | 167.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA332051000
Price (MYR)
254.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
254.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Mercaptobenzothiazole
Name Note98%
CAS149-30-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
View more
Indicator
RUO – Research Use Only