2-Decanol, 98%
2-Decanol, 98%
2-Decanol, 98%
Thermo Scientific Chemicals
2-Decanol, 98%
CAS: 1120-06-5 | C10H22O | 158.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA331600250
Price (MYR)
660.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
660.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Decanol
Name Note95%
CAS1120-06-5
CAS Min %97.5
CAS Max %100.0