5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt hydrate, 98%
CAS: 114162-64-0 | C20H26BrClN2O7 | 521.79 g/mol
Have Questions?
Change view
Quantity:
100 mg
25 mg
Packaging:
Glass bottle
Catalog number FSA331191000
Price (MYR)
1,951.00
BOT
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,951.00
BOT
Add to cart
Specifications
Chemical Name or Material5-Bromo-4-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt hydrate
Name Note98%
CAS114162-64-0
CAS Min %97.5
Health Hazard 1LIGHT SENSITIVE