Acrylamide / N,N'-Methylenebisacrylamide 37.5:1, for biochemistry, 40% mix solution in water
Acrylamide / N,N'-Methylenebisacrylamide, CAS # 79-06-1, 7732-18-5, 110-26-9, 79-06-1, is a water soluble compound that cross-links to produce polyacrylamide gels that are widely used in chromatography and the electrophoretic separation of nucleic acids and proteins. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Have Questions?
Change view
Quantity:
500 mL
100 mL
Catalog number FSA330225000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS79-06-1, 7732-18-5, 110-26-9, 79-06-1
Specifications
Absorbance(1% in water), (1% in water) at 0nm, 0.1 max. at 290nm
Chemical Name or MaterialAcrylamide / N, N'-Methylenebisacrylamide 37.5:1
Physical FormLiquid
Acrylic Acid0.001% max.
CAS79-06-1
View more
General description
Acrylamide / N,N'-Methylenebisacrylamide is a water-soluble, odorless, white crystalline powder
This compound polymerizes in the presence of free radicals to generate gels whose pore size may be regulated by manipulating the relative concentrations of this mixture of compounds
Applications
Polyacrylamide gels are widely used in biochemistry, forensic chemistry, genetics, molecular biology, and biotechnology to separate proteins or nucleic acids, according to their electrophoretic mobility
Because of their higher resolution compared to other gels, polyacrylamide gels are also used in the analysis of smaller RNA species from mixed samples