D(+)-Maltose monohydrate, 92+%
D(+)-Maltose monohydrate, 92+%
D(+)-Maltose monohydrate, 92+%
D(+)-Maltose monohydrate, 92+%
Thermo Scientific Chemicals
D(+)-Maltose monohydrate, 92+%
CAS: 6363-53-7 | C12H24O12 | 360.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA329915000
Price (MYR)
647.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
647.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD(+)-Maltose monohydrate
Name Note90%
CAS6363-53-7
Melting Point130.0°C