Brij™ L23-SO-(AP), 30 w/v % sol. in water, previous name Brij™ 35
CAS: 9002-92-0 | C58H118O24
Have Questions?
Change view
Quantity:
100 mL
1 L
2.5 L
Catalog number FSA329581000
Price (MYR)
340.00
BOT
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
340.00
BOT
Add to cart
Chemical Identifiers
CAS9002-92-0, 7732-18-5
Specifications
Chemical Name or MaterialBrij™ 35
ColorColorless
CAS9002-92-0
Physical FormJelly Substance at 25°C
Health Hazard 2GHS H Statement: May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.