Diltiazem hydrochloride, 98%
Diltiazem hydrochloride, 98%
Diltiazem hydrochloride, 98%
Diltiazem hydrochloride, 98%
Thermo Scientific Chemicals
Diltiazem hydrochloride, 98%
CAS: 33286-22-5 | C22H26N2O4S·HCl | 450.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3290800101 gGlass bottle
Catalog number FSA329080010
Price (MYR)
478.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
478.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiltiazem hydrochloride
CAS33286-22-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement