ODQ, 98%
ODQ, 98%
ODQ, 98%
Thermo Scientific Chemicals
ODQ, 98%
CAS: 41443-28-1 | C9H5N3O2 | 187.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
Packaging:
Glass bottle
Catalog number FSA328800100
Price (MYR)
989.00
EA
Add to cart
Quantity:
10 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
989.00
EA
Add to cart
Specifications
Chemical Name or MaterialODQ
CAS41443-28-1
CAS Min %97.5
CAS Max %100.0
Melting Point160°C to 170°C