NADPH tetrasodium salt hydrate, 93%, extra pure, Thermo Scientific Chemicals
NADPH tetrasodium salt hydrate, 93%, extra pure, Thermo Scientific Chemicals
NADPH tetrasodium salt hydrate, 93%, extra pure, Thermo Scientific Chemicals
NADPH tetrasodium salt hydrate, 93%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
NADPH tetrasodium salt hydrate, 93%, extra pure, Thermo Scientific Chemicals
CAS: 2646-71-1 | C21H26N7Na4O17P3 | 833.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA328742500
Price (MYR)
2,439.00
BOT
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,439.00
BOT
Add to cart
Specifications
Chemical Name or MaterialNADPH tetrasodium salt hydrate
Name Noteapprox. 98%
CAS2646-71-1
CAS Min %97.5
Health Hazard 1LIGHT SENSITIVE