Griess Reagent
Griess Reagent
Griess Reagent
Thermo Scientific Chemicals
Griess Reagent
Have Questions?
Catalog NumberQuantity
FSA32867050050 mL
Catalog number FSA328670500
Price (MYR)
940.00
BOT
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (MYR)
940.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGriess Reagent
ColorColorless to Pink
Health Hazard 1Danger
Health Hazard 2GHS P Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.