Leupeptin, 90%, synthetic
Leupeptin, 90%, synthetic
Leupeptin, 90%, synthetic
Leupeptin, 90%, synthetic
Thermo Scientific Chemicals
Leupeptin, 90%, synthetic
CAS: 103476-89-7 | 0·5 H2O4S
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
5 mg
Catalog number FSA328350250
Price (MYR)
1,554.00
EA
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (MYR)
1,554.00
EA
Add to cart
Specifications
Chemical Name or MaterialLeupeptin
ColorWhite
Product TypeLeupeptin
CAS103476-89-7
Molecular Formula0·5 H2O4S