Genistein, 99%, synthetic
Genistein, 99%, synthetic
Genistein, 99%, synthetic
Genistein, 99%, synthetic
Thermo Scientific Chemicals
Genistein, 99%, synthetic
CAS: 446-72-0 | C15H10O5 | 270.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
Packaging:
Glass bottle
Catalog number FSA328271000
Price (MYR)
1,850.00
EA
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,850.00
EA
Add to cart
Specifications
Chemical Name or MaterialGenistein
Name NoteSynthetic
CAS446-72-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement