Fumonisin B2, CAS # 116355-84-1, is a diester of tricarballylic acid and is isolated from Fusarium. | CAS: 116355-84-1 | C34H59NO14·3H2O | 759.9 g/mol
Have Questions?
Catalog Number
Quantity
FSA328260010
1 mg
Catalog number FSA328260010
Price (MYR)
3,362.00
BOT
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (MYR)
3,362.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFumonisin B2
CAS116355-84-1
Health Hazard 1GHS Signal Word: Danger
Assay Percent Range96+%
Molecular FormulaC34H59NO14·3H2O
View more
General Description
Fumonisin B2 is a fumonisin mycotoxin produced by the fungus Fusarium
Fumonisin B2 is a structural analog of fumonisin B1 but it is more cytotoxic than B1 and can inhibit sphinganine-N-acetyltransferase (ceramide synthase)
Applications
In vitro and in vivo studies have shown Fumonisin B2 can induce apoptosis
It can inhibit de novo sphingolipid biosynthesis
It is also considered an Aspergillus metabolite
It shows cytotoxicity against a panel of seven rat hepatoma cell lines, and MDCK cell line
It induces hepatocyte nodule formation, a marker of cancer initiation, in rats
RUO – Research Use Only
General References:
Gelderblom, W.C.A.; Jaskiewicz, K.; Marasas, W.F.O., et al. Fumonisins-novel mycotoxins with cancer-promoting activity produced by Fusarium moniliforme. Appl. Environ. Microbiol. 1988, 54, (7), 1806-1811.
Shier, W.T.; Abbas, H.K.; and Mirocha, C.J. Toxicity of the mycotoxins fumonisins B1 and B2 and Alternaria alternata f. sp. lycopersici toxin (AAL) in cultured mammalian cells. Mycopathologia. 1991, 116, (2), 97-104.
Gelderblom, W.C.A.; Cawood, M.E., Snyman, S.D., et al. Structure-activity relationships of fumonisins in short-term carcinogenesis and cytotoxicity assays. Food Chem. Toxicol. 1993, 31, (6), 407-414.