TAPS, 99+%, for biochemistry
TAPS, 99+%, for biochemistry
TAPS, 99+%, for biochemistry
TAPS, 99+%, for biochemistry
Thermo Scientific Chemicals
TAPS, 99+%, for biochemistry
CAS: 29915-38-6 | C7H17NO6S | 243.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA327801000
Price (MYR)
685.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
685.00
BOT
Add to cart
Specifications
Absorbance0.04 max. at 260nm
Chemical Name or MaterialTAPS
Absorbance Conditions(0.1M aq. soln.)
CAS29915-38-6
Melting Point240.0°C
View more
Biological buffer
RUO – Research Use Only