Toluene, 99.85%, Extra Dry, AcroSeal™
Toluene, 99.85%, Extra Dry, AcroSeal™
Toluene, 99.85%, Extra Dry, AcroSeal™
Toluene, 99.85%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
Catalog number FSA326980010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialToluene
Name NoteExtra Dry
CAS108-88-3
Health Hazard 1Signal Word: DANGER!
Health Hazard 2GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag