Isopropanol, 99.8%, Extra Dry, AcroSeal™
Isopropanol, 99.8%, Extra Dry, AcroSeal™
Isopropanol, 99.8%, Extra Dry, AcroSeal™
Isopropanol, 99.8%, Extra Dry, AcroSeal™
Isopropanol, 99.8%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Isopropanol, 99.8%, Extra Dry, AcroSeal™
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number FSA326961000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
CAS67-63-0
ColorColorless
Health Hazard 1Danger