Dichloromethane, for residue and pestic. anal., stab. with amylene
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number FSA326600025
Price (MYR)
444.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
444.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDichloromethane
Melting Point-97.0°C
Name NoteECD Tested, For Pesticide analysis
CAS75-09-2
Health Hazard 1Warning